MilliporeSigma
  • Rapid purification of plasmid DNA by a single centrifugation in a two-step cesium chloride-ethidium bromide gradient.
Rapid purification of plasmid DNA by a single centrifugation in a two-step cesium chloride-ethidium bromide gradient.
Biochemical and biophysical research communications (1983-12-28)
S J Garger, O M Griffith, L K Grill
ABSTRACT
A procedure for rapid, preparative purification of plasmid DNA is described and compared with a conventional equilibrium centrifugation method. A discontinuous, two-step CsCl-ethidium bromide gradient is used, with the starting position of the plasmid-containing extract being at the bottom of the tube. During centrifugation in a fixed angle rotor, covalently closed circular plasmid DNA is separated from contaminating protein, RNA, and chromosomal DNA in 5 hr. Plasmids purified by this method are considerably less contaminated with RNA than when purified by a 48-hr equilibrium run in a homogeneous gradient, as determined by agarose gel electrophoresis and 5'-end-labeling studies. Plasmid DNA purified in two-step gradients can be used directly for restriction endonuclease analysis and DNA sequencing.
Sigma-Aldrich
Cesium chloride, optical grade, ≥99.5% trace metals basis