Merck
  • Vitronectin expression and interaction with receptors in smooth muscle cells from human atheromatous plaque.
Vitronectin expression and interaction with receptors in smooth muscle cells from human atheromatous plaque.
Arteriosclerosis, thrombosis, and vascular biology (1998-03-04)
P Dufourcq, H Louis, C Moreau, D Daret, M R Boisseau, J M Lamazière, J Bonnet
ABSTRACT
Vitronectin (VN) is a plasma glycoprotein that promotes cell attachment and induces migration of human smooth muscle cells (SMCs) in culture. VN has been observed to accumulate in human atherosclerotic plaques, although its origin and role in atherosclerosis are not yet established. In the present experiments, synthesis of VN by intimal cells and its colocalization with receptors, alphavbeta3 and alphavbeta5, were studied by in situ hybridization and immunohistochemistry on 15 human atherosclerotic plaques from carotid arteries obtained after surgery. Strong VN protein and mRNA expression was observed in the intima and in the media. In the intima, VN mRNA expression was colocalized with SMCs, indicating that these cells produce VN, which may account for its accumulation in atherosclerotic plaques. In SMCs in culture, immunoprecipitation after metabolic labeling demonstrated that human SMCs do synthesize vitronectin. Confocal microscopic examination showed that VN colocalized with its receptors, alphavbeta3 and alphavbeta5, in the atherosclerotic intima. However, the distribution of the VN receptors on SMCs in culture in contact with VN was different. These observations suggest that VN plays various parts in atherogenesis via different SMC membrane receptors.
Sigma-Aldrich
Monoclonal Anti-Vitronectin antibody produced in mouse, clone VIT-2, ascites fluid