MilliporeSigma
Inhibition of H3K27me3 Demethylases Promotes Plasmablast Formation.
ImmunoHorizons (2021-12-10)
Anna K Kania, Muyao Guo, Christopher D Scharer, Jeremy M Boss
ABSTRACT
B cell differentiation into Ab-secreting plasma cells requires transcriptional, metabolic, and epigenetic remodeling. Histone H3 lysine 27 trimethylation (H3K27me3), a histone modification associated with gene silencing, is dynamically regulated during B cell differentiation. Although several studies have focused on mechanisms involving the gain of this modification in plasmablasts (PB), the role of active demethylation of H3K27me3 by ubiquitously transcribed tetratricopeptide repeat, X chromosome (UTX) and Jumonji domain-containing protein 3 (JMDJ3) during B cell differentiation has not been examined. In this study, this process was assessed using a pharmacological inhibitor of UTX and JMJD3, GSK-J4. Treatment of ex vivo stimulated mouse B cells with GSK-J4 led to an increase in PB frequency without affecting the ability of the newly formed PB to secrete Abs. Consistent with the role of UTX and JMJD3 in promoting gene expression, the majority of differentially expressed were downregulated upon GSK-J4 treatment. GSK-J4-treated cells downregulated genes associated with signaling and P53 pathways. Inhibitor treated cells upregulated genes associated with cell cycle and proliferation, which correlated with an increase in actively proliferating cells. Unexpectedly, a majority of the downregulated transcripts corresponded to genes that in the wild-type setting were genes that gain H3K27me3 and downregulated in PB. Together, our results show that UTX and JMDJ3 are required to restrain B cell differentiation and suggest that they function as a rheostat for H3K27me3 to control this process.
Sigma-Aldrich
GSK-J4, ≥98% (HPLC)