MilliporeSigma
  • Global fitting for high-accuracy multi-channel single-molecule localization.
Global fitting for high-accuracy multi-channel single-molecule localization.
Nature communications (2022-06-07)
Yiming Li, Wei Shi, Sheng Liu, Ivana Cavka, Yu-Le Wu, Ulf Matti, Decheng Wu, Simone Koehler, Jonas Ries
ABSTRACT
Multi-channel detection in single-molecule localization microscopy greatly increases information content for various biological applications. Here, we present globLoc, a graphics processing unit based global fitting algorithm with flexible PSF modeling and parameter sharing, to extract maximum information from multi-channel single molecule data. As signals in multi-channel data are highly correlated, globLoc links parameters such as 3D coordinates or photon counts across channels, improving localization precision and robustness. We show, both in simulations and experiments, that global fitting can substantially improve the 3D localization precision for biplane and 4Pi single-molecule localization microscopy and color assignment for ratiometric multicolor imaging.
Sigma-Aldrich
Anti-AHCTF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution