Merck
  • Complexation with a Cognate Antibody Fragment Facilitates Affinity Measurements of Fluorescein-Linked Small Molecule Ligands.
Complexation with a Cognate Antibody Fragment Facilitates Affinity Measurements of Fluorescein-Linked Small Molecule Ligands.
Analytical chemistry (2020-07-04)
Marco Catalano, Sebastian Oehler, Luca Prati, Nicholas Favalli, Gabriele Bassi, Jörg Scheuermann, Dario Neri
ABSTRACT
The availability of reliable methods for the characterization of the binding of small molecule ligands to protein targets is crucially important for drug discovery. We have adapted a method, routinely used for the characterization of monoclonal antibodies (enzyme-linked immunosorbent assay, or "ELISA"), to small molecule ligands, using fluorescein conjugates and antifluorescein antibodies as detection reagents. The new small molecule-ELISA methodology was tested using a panel of binders specific to carbonic anhydrase II, with dissociation constants ranging between 6 μM and 14 nM. An excellent agreement was found between ELISA measurements and fluorescence polarization results. The methodology was also extended to BIAcore measurements and implemented for ligands coupled to oligonucleotides. Small molecule-ELISA procedures are particularly useful in the context of DNA-encoded libraries, for which hit validation procedures need to be performed on dozens of candidate molecules and hit compounds can be conveniently resynthesized on DNA.
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
3, 3′,5 ,5′-Tetramethylbenzidine Liquid Substrate, Supersensitive, for ELISA, ready to use solution