MilliporeSigma
  • Novel Iron Oxide Nanoparticles Induce Ferroptosis in a Panel of Cancer Cell Lines.
Novel Iron Oxide Nanoparticles Induce Ferroptosis in a Panel of Cancer Cell Lines.
Molecules (Basel, Switzerland) (2022-07-10)
Roberto Fernández-Acosta, Claudia Iriarte-Mesa, Daniel Alvarez-Alminaque, Behrouz Hassannia, Bartosz Wiernicki, Alicia M Díaz-García, Peter Vandenabeele, Tom Vanden Berghe, Gilberto L Pardo Andreu
ABSTRACT
The use of nanomaterials rationally engineered to treat cancer is a burgeoning field that has reported great medical achievements. Iron-based polymeric nano-formulations with precisely tuned physicochemical properties are an expanding and versatile therapeutic strategy for tumor treatment. Recently, a peculiar type of regulated necrosis named ferroptosis has gained increased attention as a target for cancer therapy. Here, we show for the first time that novel iron oxide nanoparticles coated with gallic acid and polyacrylic acid (IONP-GA/PAA) possess intrinsic cytotoxic activity on various cancer cell lines. Indeed, IONP-GA/PAA treatment efficiently induces ferroptosis in glioblastoma, neuroblastoma, and fibrosarcoma cells. IONP-GA/PAA-induced ferroptosis was blocked by the canonical ferroptosis inhibitors, including deferoxamine and ciclopirox olamine (iron chelators), and ferrostatin-1, the lipophilic radical trap. These ferroptosis inhibitors also prevented the lipid hydroperoxide generation promoted by the nanoparticles. Altogether, we report on novel ferroptosis-inducing iron encapsulated nanoparticles with potent anti-cancer properties, which has promising potential for further in vivo validation.
Sigma-Aldrich
Necrostatin-1, Necrostatin-1, CAS 4311-88-0, is a cell-permeable, potent, and selective blocker of necroptosis (EC₅₀ = 494 nM in FADD-deficient Jurkat cells treated with TNF-α).