MilliporeSigma
  • Skeletal muscle fatty acid transporter protein expression in type 2 diabetes patients compared with overweight, sedentary men and age-matched, endurance-trained cyclists.
Skeletal muscle fatty acid transporter protein expression in type 2 diabetes patients compared with overweight, sedentary men and age-matched, endurance-trained cyclists.
Acta physiologica (Oxford, England) (2007-03-31)
M M A L Pelsers, K Tsintzas, H Boon, K Jewell, L Norton, J J F P Luiken, J F C Glatz, L J C van Loon
ABSTRACT
Membrane fatty acid transporters can modulate the balance between fatty acid uptake and subsequent storage and/or oxidation in muscle tissue. As such, skeletal muscle fatty acid transporter protein expression could play an important role in the etiology of insulin resistance and/or type 2 diabetes. In the present study, fatty acid translocase (FAT/CD36), plasma membrane-bound fatty acid-binding protein (FABPpm) and fatty acid transport protein 1 (FATP1) mRNA and protein expression were assessed in muscle tissue obtained from 10 sedentary, overweight type 2 diabetes patients (60 +/- 2 years), 10 sedentary, weight-matched normoglycemic controls (60 +/- 2 years) and 10 age-matched, endurance trained cyclists (57 +/- 1 years). Both FAT/CD36 and FATP1 mRNA and protein expression did not differ between groups. In contrast, FABPpm mRNA and protein expression were approx. 30-40% higher in the trained men compared with the diabetes patients (P < 0.01) and sedentary controls (P < 0.05). Skeletal muscle FAT/CD36, FABPpm and FATP1 mRNA and protein expression are not up- or downregulated in a sedentary and/or insulin resistant state. In contrast, FABPpm expression is upregulated in the endurance trained state and likely instrumental to allow greater fatty acid oxidation rates.
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Sign Into View Organizational & Contract Pricing