MilliporeSigma
  • Cell-penetrating peptides enhance the transduction of adeno-associated virus serotype 9 in the central nervous system.
Cell-penetrating peptides enhance the transduction of adeno-associated virus serotype 9 in the central nervous system.
Molecular therapy. Methods & clinical development (2021-03-27)
Yuan Meng, Dong Sun, Yiyan Qin, Xiaoyi Dong, Guangzuo Luo, Ying Liu
ABSTRACT
Recombinant adeno-associated viruses (rAAVs) have been widely used in the gene therapy field for decades. However, because of the challenge of effectively delivering rAAV vectors through the blood-brain barrier (BBB), their applications for treatment of central nervous system (CNS) diseases are quite limited. In this study, we found that several cell-penetrating peptides (CPPs) can significantly enhance the in vitro transduction efficiency of AAV serotype 9 (AAV9), a promising AAV vector for treatment of CNS diseases, the best of which was the LAH4 peptide. The enhancement of AAV9 transduction by LAH4 relied on binding of the AAV9 capsid to the peptide. Furthermore, we demonstrated that the LAH4 peptide increased the AAV9 transduction in the CNS in vitro and in vivo after systemic administration. Taken together, our results suggest that CPP peptides can interact directly with AAV9 and increase the ability of this AAV vector to cross the BBB, which further induces higher expression of target genes in the brain. Our study will help to improve the applications of AAV gene delivery vectors for the treatment of CNS diseases.
Sigma-Aldrich
Blood-Brain Barrier hCMEC/D3 Cell Line, The hCMEC/D3 BBB cell line has been extensively characterized for brain endothelial phenotype and is a model of human blood-brain barrier (BBB) function.