MilliporeSigma
  • Impact of maternal intermittent fasting during pregnancy on cardiovascular, metabolic and renal function in adult rat offspring.
Impact of maternal intermittent fasting during pregnancy on cardiovascular, metabolic and renal function in adult rat offspring.
PloS one (2022-03-11)
Alaa Alkhalefah, Heather J Eyre, Rezwana Hussain, Jocelyn D Glazier, Nick Ashton
ABSTRACT
Pregnant Muslim women are exempt from fasting during Ramadan; however a majority are reported to fast. The impact of this form of maternal intermittent fasting (IF) on fetal development and offspring health is not well defined. Using a rat model, we have shown previously that maternal IF results in fetal growth restriction accompanied by changes in placental nutrient transport function. The aim of this study was to assess cardiovascular, metabolic and renal function in adult offspring of IF-exposed dams. Food was withheld from Wistar rats from 17:00 to 09:00 daily throughout pregnancy; controls had ad libitum access to food. Birth weight was unaffected; however male IF pups grew more slowly up to 10 weeks of age (P < 0.01) whereas IF females matched their control counterparts. Systolic blood pressure (SBP), glucose tolerance and basal renal function at 14 weeks were not affected by IF exposure. When offered saline solutions (0.9-2.1%) to drink, females showed a greater salt preference than males (P < 0.01); however there were no differences between dietary groups. A separate group of pups was weaned onto a 4% NaCl diet. SBP increased in IF pups sooner, at 7 weeks (P < 0.01), than controls which became hypertensive from 10 weeks. Renal function did not appear to differ; however markers of renal injury were elevated in IF males (P < 0.05). Maternal IF does not affect resting cardiovascular, metabolic and renal function; but when challenged by dietary salt load male IF offspring are more prone to renal injury.
Sigma-Aldrich
Rat Lcn2 / Neutrophil Gelatinase-associated Lipocalin ELISA Kit
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture