MilliporeSigma
  • AIE-active polyanetholesulfonic acid sodium salts with room-temperature phosphorescence characteristics for Fe3+ detection.
AIE-active polyanetholesulfonic acid sodium salts with room-temperature phosphorescence characteristics for Fe3+ detection.
RSC advances (2018-09-04)
Xiaojin Qin, Shiqi Wang, Lian Luo, Gang He, Huazhen Sun, Yongyang Gong, Bingli Jiang, Chun Wei
ABSTRACT
Room-temperature phosphorescent materials have been a major focus of research and development during the past decades, due to their applications in OLEDs, photovoltaic cells, chemical sensors, and bioimaging. However, achieving polymeric phosphorescent materials without heavy-metal atoms and halogens under ambient conditions remains a major challenge. Here, we report a polymeric phosphor, namely polyanetholesulfonic acid sodium salt, which not only has room temperature phosphorescence characteristic but also aggregation-induced emission and dependence on the excitation wavelength characteristics. Moreover, it can recognize Fe3+ effectively.
Sigma-Aldrich
Polyanetholesulfonic acid sodium salt, powder