MilliporeSigma
  • Alterations of beta-catenin pathway in non-melanoma skin tumors: loss of alpha-ABC nuclear reactivity correlates with the presence of beta-catenin gene mutation.
Alterations of beta-catenin pathway in non-melanoma skin tumors: loss of alpha-ABC nuclear reactivity correlates with the presence of beta-catenin gene mutation.
The American journal of pathology (2003-11-25)
Claudio Doglioni, Sara Piccinin, Silvia Demontis, Maria Giulia Cangi, Lorenza Pecciarini, Concetta Chiarelli, Michela Armellin, Tamara Vukosavljevic, Mauro Boiocchi, Roberta Maestro
ABSTRACT
To determine the role of beta-catenin pathway in human skin carcinogenesis, 135 non-melanoma skin tumors were analyzed for beta-catenin expression and gene mutations. Intense nucleo-cytoplasmic immunoreactivity for C terminus beta-catenin antibodies was observed in all pilomatricomas and in single cases of trichoepithelioma and squamous cell carcinoma showing peculiar signs of matrical differentiation. Moderate increase of beta-catenin nuclear staining was detected in a significant proportion of basal cell carcinomas, Bowen disease, spiroadenomas, and occasionally also in squamous cell carcinomas, but in these neoplasms only a limited fraction of tumor cells accumulated beta-catenin. Molecular analysis revealed that beta-catenin gene mutations are a peculiar feature of skin tumors with matrical differentiation and correlate with a pattern of intense and diffuse beta-catenin nuclear expression. In contrast, adenomatous polyposis coli (APC) and AXIN2 mutations were not involved in skin tumorigenesis. Analysis of Wnt pathway revealed that TCF-1 and MITF-M were selectively induced in the tumor types harboring beta-catenin mutations, indicating that a Wnt/beta-catenin pathway involving TCF-1 and MITF-M is activated in these tumors. Interestingly, high expression levels of TCF-3 were found in basal cell carcinomas and spiroadenomas. TCF-3 is reported to act as a negative modulator of beta-catenin degradation pathway. Thus, the moderate increase of beta-catenin nuclear staining detected in these tumor types might, at least in part, be due to a TCF-3-dependent mechanism. Finally, we found that the presence of beta-catenin mutations significantly correlated with loss of nuclear immunoreactivity for an antibody raised against the N terminus of beta-catenin (alphaABC). Thus, a combined analysis with C terminus-beta-catenin antibodies and alphaABC Ab may represent a powerful investigative approach for the detection of beta-catenin structural alterations.
Sigma-Aldrich
Monoclonal Anti-β-Catenin antibody produced in mouse, clone 15B8, ascites fluid
Sigma-Aldrich
Anti-TCF-4 Antibody, clone 6H5-3, clone 6H5-3, Upstate®, from mouse