MilliporeSigma
  • Antiviral Effect of hBD-3 and LL-37 during Human Primary Keratinocyte Infection with West Nile Virus.
Antiviral Effect of hBD-3 and LL-37 during Human Primary Keratinocyte Infection with West Nile Virus.
Viruses (2022-07-28)
Céline Chessa, Charles Bodet, Clément Jousselin, Andy Larivière, Alexia Damour, Julien Garnier, Nicolas Lévêque, Magali Garcia
ABSTRACT
West Nile virus (WNV) is an emerging flavivirus transmitted through mosquito bites and responsible for a wide range of clinical manifestations. Following their inoculation within the skin, flaviviruses replicate in keratinocytes of the epidermis, inducing an innate immune response including the production of antimicrobial peptides (AMPs). Among them, the cathelicidin LL-37 and the human beta-defensin (hBD)-3 are known for their antimicrobial and immunomodulatory properties. We assessed their role during WNV infection of human primary keratinocytes. LL-37 reduced the viral load in the supernatant of infected keratinocytes and of the titer of a viral inoculum incubated in the presence of the peptide, suggesting a direct antiviral effect of this AMP. Conversely, WNV replication was not inhibited by hBD-3. The two peptides then demonstrated immunomodulatory properties whether in the context of keratinocyte stimulation by poly(I:C) or infection by WNV, but not alone. This study demonstrates the immunostimulatory properties of these two skin AMPs at the initial site of WNV replication and the ability of LL-37 to directly inactivate West Nile viral infectious particles. The results provide new information on the multiple functions of these two peptides and underline the potential of AMPs as new antiviral strategies in the fight against flaviviral infections.
Sigma-Aldrich
Anti-Viperin Antibody, clone MaP.VIP, clone MaP.VIP, from mouse
Roche
Cell Proliferation Kit II (XTT), liquid, pkg of 1 kit, suitable for cell analysis, suitable for tissue culture
Sigma-Aldrich
bisBenzimide H 33258, powder, BioReagent, suitable for cell culture, ≥98% (HPLC and TLC)