Merck
  • High-yield overproduction and purification of human aquaporins from Pichia pastoris.
High-yield overproduction and purification of human aquaporins from Pichia pastoris.
STAR protocols (2022-04-26)
Tamim Al-Jubair, Jonas Hyld Steffen, Julie Winkel Missel, Philip Kitchen, Mootaz M Salman, Roslyn M Bill, Pontus Gourdon, Susanna Törnroth-Horsefield
ABSTRACT
Aquaporins (AQPs) are membrane-bound water channels that play crucial roles in maintaining the water homeostasis of the human body. Here, we present a protocol for high-yield recombinant expression of human AQPs in the methylotropic yeast Pichia pastoris and subsequent AQP purification. The protocol typically yields 1-5 mg AQP per g of yeast cell at >95% purity and is compatible with any membrane protein cloned into Pichia pastoris, although expression levels may vary. For complete details on the use and execution of this protocol, please refer to Kitchen et al. (2020) and Frick et al. (2014).
Sigma-Aldrich
Select Yeast Extract, powder, BioReagent, suitable for cell culture, suitable for insect cell culture