Merck
  • IGF-1 Receptor Signaling Regulates Type II Pneumocyte Senescence and Resulting Macrophage Polarization in Lung Fibrosis.
IGF-1 Receptor Signaling Regulates Type II Pneumocyte Senescence and Resulting Macrophage Polarization in Lung Fibrosis.
International journal of radiation oncology, biology, physics (2021-01-02)
Eun Joo Chung, Seokjoo Kwon, Jessica L Reedy, Ayla O White, Joon Seon Song, Ilseon Hwang, Joon Yong Chung, Kris Ylaya, Stephen M Hewitt, Deborah E Citrin
ABSTRACT
Type II pneumocyte (alveolar epithelial cells type II [AECII]) senescence has been implicated in the progression of lung fibrosis. The capacity of senescent cells to modulate pulmonary macrophages to drive fibrosis is unexplored. Insulin-like growth factor-1 receptor (IGF-1R) signaling has been implicated as a regulator of senescence and aging. Mice with an AECII-specific deletion of IGF-1R received thoracic irradiation (n ≥ 5 per condition), and the effect of IGF-1R deficiency on radiation-induced AECII senescence and macrophage polarization to an alternatively activated phenotype (M2) was investigated. IGF-1R signaling, macrophage polarization, and senescence were evaluated in surgically resected human lung (n = 63). IGF-1R deficient mice demonstrated reduced AECII senescence (senescent AECII/field; intact: 7.25% ± 3.5% [mean ± SD], deficient: 2.75% ± 2.8%, P = .0001), reduced accumulation of M2 macrophages (intact: 24.7 ± 2.2 cells/field, deficient: 15.5 ± 1.2 cells/field, P = .0086), and fibrosis (hydroxyproline content; intact: 71.9 ± 21.7 μg/lung, deficient: 31.7 ± 7.9, P = .0485) after irradiation. Senescent AECII enhanced M2 polarization in a paracrine fashion (relative Arg1 mRNA, 0 Gy: 1.0 ± 0.4, 17.5 Gy: 7.34 ± 0.5, P < .0001). Evaluation of surgical samples from patients treated with chemoradiation demonstrated increased expression of IGF-1 (unirradiated: 10.2% ± 4.9% area, irradiated: 15.1% ± 11.5%, P = .0377), p21 (unirradiated: 0.013 ± 0.02 histoscore, irradiated: 0.084 ± 0.09 histoscore, P = .0002), IL-13 (unirradiated: 13.7% ± 2.8% area, irradiated: 21.7% ± 3.8%, P < .0001), and M2 macrophages in fibrotic regions relative to nonfibrotic regions (unirradiated: 11.4 ± 12.2 CD163 + cells/core, irradiated: 43.1 ± 40.9 cells/core, P = .0011), consistent with findings from animal models of lung fibrosis. This study demonstrates that senescent AECII are necessary for the progression of pulmonary fibrosis and serve as a targetable, chronic stimuli for macrophage activation in fibrotic lung.
Sigma-Aldrich
Anti-IGF1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.