Merck
Effect of α-Amylase on the Structure of Chia Seed Mucilage.
Biomimetics (Basel, Switzerland) (2022-10-25)
Francesco Piazza, Matilde Colella, Giuseppe Cinelli, Francesco Lopez, Ivan Donati, Pasquale Sacco
ABSTRACT
Thanks to its nutritional and mechanical properties, chia seed mucilage is becoming increasingly popular in the food industry as a small biomolecule. The mechanical properties of an ingredient are a key element for food appreciation during chewing. Therefore, with this study, we explore for the first time the structural changes that chia seed mucilage undergoes when treated with α-amylase, the most abundant enzyme in human saliva. First, rheological time-sweep tests were performed on samples with different enzyme and constant chia mucilage concentrations. Then, the effect of increasing the chia mucilage concentration at a constant enzyme content was investigated. The results show that structural changes occur after enzyme treatment. Rheological measurements show a thickening of the material with an increase in the elastic modulus depending on the concentrations of α-amylase and chia used. This effect is attributed to the release and aggregation of insoluble fibrous aggregates that naturally form the mucilage after the cleavage of the α-1,4-glucoside bond between the α-D-glucopyranose residue and the second β-D-xylopyranose residue by α-amylase. Thus, our data suggest an α-amylase-mediated restructuring of the chia mucilage network that could have implications for the commercial processing of this material.
Sigma-Aldrich
α-Amylase from Bacillus sp., powder, yellow-brown, ~50 U/mg