MilliporeSigma
  • Novel localizations of TRPC5 channels suggest novel and unexplored roles: A study in the chick embryo brain.
Novel localizations of TRPC5 channels suggest novel and unexplored roles: A study in the chick embryo brain.
Developmental neurobiology (2021-10-28)
Sharifuddin Rifat Ahmed, Elise Liu, Alissa Yip, Yuqi Lin, Evan Balaban, Maria Pompeiano
ABSTRACT
Mammalian TRPC5 channels are predominantly expressed in the brain, where they increase intracellular Ca2+ and induce depolarization. Because they augment presynaptic vesicle release, cause persistent neural activity, and show constitutive activity, TRPC5s could play a functional role in late developmental brain events. We used immunohistochemistry to examine TRPC5 in the chick embryo brain between 8 and 20 days of incubation, and provide the first detailed description of their distribution in birds and in the whole brain of any animal species. Stained areas substantially increased between E8 and E16, and staining intensity in many areas peaked at E16, a time when chick brains first show organized patterns of whole-brain metabolic activation like what is seen consistently after hatching. Areas showing cell soma staining match areas showing Trpc5 mRNA or protein in adult rodents (cerebral cortex, hippocampus, amygdala, cerebellar Purkinje cells). Chick embryos show protein staining in the optic tectum, cerebellar nuclei, and several brainstem nuclei; equivalent areas in the Allen Institute mouse maps express Trpc5 mRNA. The strongest cell soma staining was found in a dorsal hypothalamic area (matching a group of parvicellular arginine vasotocin neurons and a pallial amygdalohypothalamic cell corridor) and the vagal motor complex. Purkinje cells showed strong dendritic staining at E20. Unexpectedly, we also describe neurite staining in the septum, several hypothalamic nuclei, and a paramedian raphe area; the strongest neurite staining was in the median eminence. These novel localizations suggest new unexplored TRPC5 functions, and possible roles in late embryonic brain development.
Sigma-Aldrich
Anti-Tryptophan Hydroxylase Antibody, Chemicon®, from sheep
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Choline Acetyltransferase Antibody, Chemicon®, from goat