MilliporeSigma
  • Microtubule-dependent changes in morphology and localization of chloride transport proteins in gill mitochondria-rich cells of the tilapia, Oreochromis mossambicus.
Microtubule-dependent changes in morphology and localization of chloride transport proteins in gill mitochondria-rich cells of the tilapia, Oreochromis mossambicus.
Journal of morphology (2016-05-31)
Wen-Kai Yang, Yu-Ching Wu, Cheng-Hao Tang, Tsung-Han Lee
ABSTRACT
The tilapia (Oreochromis mossambicus) is a euryhaline fish exhibiting adaptive changes in cell size, phenotype, and ionoregulatory functions upon salinity challenge. Na(+) /Cl(-) cotransporter (NCC) and Na(+) /K(+) /2Cl(-) cotransporter (NKCC) are localized in the apical and basolateral membranes of mitochondria-rich (MR) cells of the gills. These cells are responsible for chloride absorption (NCC) and secretion (NKCC), respectively, thus, the switch of gill NCC and NKCC expression is a crucial regulatory mechanism for salinity adaptation in tilapia. However, little is known about the interaction of cytoskeleton and these adaptive changes. In this study, we examined the time-course of changes in the localization of NKCC/NCC in the gills of tilapia transferred from fresh water (FW) to brackish water (20‰) and from seawater (SW; 35‰) to FW. The results showed that basolateral NKCC disappeared and NCC was expressed in the apical membrane of MR cells. To further clarify the process of these adaptive changes, colchicine, a specific inhibitor of microtubule-dependent cellular regulating processes was used. SW-acclimated tilapia were transferred to SW, FW, and FW with colchicine (colchicine-FW) for 96 h. Compared with the FW-treatment group, in the MR cells of colchicine-FW-treatment group, (1) the average size was significantly larger, (2) only wavy-convex-subtype apical surfaces were found, and (3) the basolateral (cytoplasmic) NKCC signals were still exhibited. Taken together, our results suggest that changes in size, phenotype, as well as the expression of NCC and NKCC cotransporters of MR cells in the tilapia are microtubule-dependent. J. Morphol. 277:1113-1122, 2016. © 2016 Wiley Periodicals, Inc.
Sigma-Aldrich
Colchicine, ≥95% (HPLC), powder
Pricing and availability is not currently available.