MilliporeSigma
  • An efficient synthesis of trace amine-associated receptor agonist [1-14 C]tyramine.
An efficient synthesis of trace amine-associated receptor agonist [1-14 C]tyramine.
Journal of labelled compounds & radiopharmaceuticals (2018-07-15)
Crist N Filer, Demetri Orphanos
ABSTRACT
The 2-step synthesis of [1-14 C]tyramine hydrochloride is described with the product being characterized by TLC, HPLC, and UV spectroscopy. Several methods are provided to purify [1-14 C]tyramine hydrochloride, and its storage and stability are also discussed.
Sigma-Aldrich
Tyramine hydrochloride, ≥98%