Merck
  • Borneol protects against cerulein-induced oxidative stress and inflammation in acute pancreatitis mice model.
Borneol protects against cerulein-induced oxidative stress and inflammation in acute pancreatitis mice model.
Environmental toxicology (2020-11-10)
Sapana Bansod, Shrilekha Chilvery, Mohd Aslam Saifi, Tridip Jyoti Das, Hui Tag, Chandraiah Godugu
ABSTRACT
Borneol is a commonly used flavouring substance in traditional Chinese medicine, which possesses several pharmacological activities including analgesic, antiinflammatory, and antioxidant properties. The aim of this study was to investigate the effects of borneol on cerulein-induced acute pancreatitis (AP) model. Swiss albino mice were pretreated with borneol (100 and 300 mg/kg) daily for 7 days, before six consecutive injections of cerulein (50 μg/kg/hr, intraperitoneally). The protective effect of borneol was studied by biochemical, enzyme linked immunosorbent assay, histological, immunoblotting, and immunohistochemical analysis. Oral administration of borneol significantly attenuated pancreatic damage by reducing amylase, lipase levels and histological changes. Borneol attenuated cerulein-induced oxidative-nitrosative stress by decreasing malondialdehyde, nitrite levels, and elevating reduced glutathione levels. Pancreatic inflammation was ameliorated by inhibiting myeloperoxidase activity and pro-inflammatory cytokine (Interleukins and TNF-α) levels. Furthermore, borneol administration significantly increased nuclear factor E2-related factor 2 (Nrf2), superoxide dismutase (SOD1) expression and reduced phospho-NF-κB p65 expression. Treatment with borneol significantly inhibited TNF-α, IL-1β, IL-6, and inducible nitric oxide synthase expression in cerulein-induced AP mouse model. Together, these results indicate that borneol which is currently used as US-FDA approved food adjuvant has the potential to attenuate cerulein-induced AP possibly by reducing the oxidative damage and pancreatic inflammation by modulating Nrf2/NF-κB pathway.
Sigma-Aldrich
Monoclonal Anti-Nitric Oxide Synthase, Inducible antibody produced in mouse, clone NOS-IN, ascites fluid