- Cooling-activated dorsomedial hypothalamic BDNF neurons control cold defense in mice.
Cooling-activated dorsomedial hypothalamic BDNF neurons control cold defense in mice.
BDNF and its expressing neurons in the brain critically control feeding and energy expenditure (EE) in both rodents and humans. However, whether BDNF neurons would function in thermoregulation during temperature challenges is unclear. Here, we show that BDNF neurons in the dorsomedial hypothalamus (DMHBDNF ) of mice are activated by afferent cooling signals. These cooling-activated BDNF neurons are mainly GABAergic. Activation of DMHBDNF neurons or the GABAergic subpopulations is sufficient to increase body temperature, EE, and physical activity. Conversely, blocking DMHBDNF neurons substantially impairs cold defense and reduces energy expenditure, physical activity, and UCP1 expression in BAT, which eventually results in bodyweight gain and glucose/insulin intolerance. Therefore, we identify a subset of DMHBDNF neurons as a novel type of cooling-activated neurons to promote cold defense. Thus, we reveal a critical role of BDNF circuitry in thermoregulation, which deepens our understanding of BDNF in controlling energy homeostasis and obesity.