MilliporeSigma
  • Motor nerve damage is associated with anti-ganglioside antibodies in diabetes.
Motor nerve damage is associated with anti-ganglioside antibodies in diabetes.
Journal of the peripheral nervous system : JPNS (2004-09-15)
Sabrina Matà, Elisabetta Betti, Giulio Masotti, Francesco Pinto, Francesco Lolli
ABSTRACT
Few reports exist on the association between the humoral immune response to glycolipids and neuropathic findings in diabetes. To address this issue, we assayed serum anti-GM1, GD1b, GD1a, and sulfatides IgG and IgM in a group of 85 non-selected diabetic patients, and correlated these antibodies to clinical and electrophysiological findings. Diabetic patients had higher mean titers of anti-GM1 (IgM), GD1b, GD1a, and sulfatide (IgG) antibodies when compared to healthy controls. Patients with a positive titer of anti-ganglioside antibodies had significant alterations of motor conduction parameters. The statistical significance increased when considering those patients with both anti-ganglioside reactivity and a high value for glycosylated hemoglobin. Production of antibodies to ganglioside may follow the exposure of hidden motor nerve epitopes in damaged motor nerves and contribute to the neuropathy.
Sigma-Aldrich
Anti-Human IgG (γ-chain specific), F(ab′)2 fragment−Alkaline Phosphatase antibody produced in goat, affinity isolated antibody, buffered aqueous glycerol solution