MilliporeSigma
  • Anti-Diabetic Agent Sodium Tungstate Induces the Secretion of Pro- and Anti-Inflammatory Cytokines by Human Kidney Cells.
Anti-Diabetic Agent Sodium Tungstate Induces the Secretion of Pro- and Anti-Inflammatory Cytokines by Human Kidney Cells.
Journal of cellular physiology (2016-10-31)
Romina Bertinat, Francisco Westermeier, Pamela Silva, Jie Shi, Francisco Nualart, Xuhang Li, Alejandro J Yáñez
ABSTRACT
Diabetic kidney disease (DKD) is the major cause of end stage renal disease. Sodium tungstate (NaW) exerts anti-diabetic and immunomodulatory activities in diabetic animal models. Here, we used primary cultures of renal proximal tubule epithelial cells derived from type-2-diabetic (D-RPTEC) and non-diabetic (N-RPTEC) subjects as in vitro models to study the effects of NaW on cytokine secretion, as these factors participate in intercellular regulation of inflammation, cell growth and death, differentiation, angiogenesis, development, and repair, all processes that are dysregulated during DKD. In basal conditions, D-RPTEC cells secreted higher levels of prototypical pro-inflammatory IL-6, IL-8, and MCP-1 than N-RPTEC cells, in agreement with their diabetic phenotype. Unexpectedly, NaW further induced IL-6, IL-8, and MCP-1 secretion in both N- and D-RPTEC, together with lower levels of IL-1 RA, IL-4, IL-10, and GM-CSF, suggesting that it may contribute to the extent of renal damage/repair during DKD. Besides, NaW induced the accumulation of IκBα, the main inhibitor protein of one major pathway involved in cytokine production, suggesting further anti-inflammatory effect in the long-term. A better understanding of the mechanisms involved in the interplay between the anti-diabetic and immunomodulatory properties of NaW will facilitate future studies about its clinical relevance. J. Cell. Physiol. 232: 355-362, 2017. © 2016 Wiley Periodicals, Inc.
Sigma-Aldrich
MKK6/SKK3 Protein, inactive, 50 g, Unactive, recombinant human MKK6, amino acids 4-end, fused at the N-terminus to a Mal-E tag, for use in Kinase Assays.