Merck
  • BMI1 induces ubiquitination and protein degradation of Nod-like receptor family CARD domain containing 5 and suppresses human leukocyte antigen class I expression to induce immune escape in non-small cell lung cancer.
BMI1 induces ubiquitination and protein degradation of Nod-like receptor family CARD domain containing 5 and suppresses human leukocyte antigen class I expression to induce immune escape in non-small cell lung cancer.
The Kaohsiung journal of medical sciences (2022-10-05)
Zhi-Hui Lu, Gan-Jie Tu, Si-Lv Fu, Kai Shang, Su-Juan Peng, Li Chen, Xi-Juan Gu
ABSTRACT
The Nod-like receptor (NLR) family CARD domain containing 5 (NLRC5) has been reported as an activator of human leukocyte antigen (HLA) class I that is responsible for immune activity in cancer treatment. This work focuses on the role of BMI1 proto-oncogene (BMI1) in the NLRC5-HLA class I axis and in immune escape in non-small cell lung cancer (NSCLC). First, immunoblot analysis and/or reverse transcription-quantitative polymerase chain reaction were performed, which identified decreased NLRC5 and HLA class I levels in NSCLC tissues and cell lines. NSCLCs were co-cultured with activated CD8+ T cells. Overexpression of NLRC5 in NSCLC cells elevated the expression of HLA class I and increased the activity of T cells and IL-2 production, and it reduced the PD-1/PD-L1 levels. The ubiquitination and immunoprecipitation assays confirmed that BMI1 bound to NLRC5 to induce is ubiquitination and protein degradation. Downregulation of BMI1 in NSCLC cells elevated NLRC5 and HLA class I levels, and consequently promoted T cell activation and decreased PD-1/PD-L1 levels in the co-culture system. However, overexpression of BMI1 in cells led to inverse trends. In summary, this study demonstrates that BMI1 induces ubiquitination and protein degradation of NLRC5 and suppresses HLA class I expression, which potentially helps immune escape in NSCLC.
Sigma-Aldrich
Anti-HA Magnetic Beads, Magnetic agarose, suspension