MilliporeSigma
  • The miR-124-3p regulates the allergic airway inflammation and remodeling in an ovalbumin-asthmatic mouse model by inhibiting S100A4.
The miR-124-3p regulates the allergic airway inflammation and remodeling in an ovalbumin-asthmatic mouse model by inhibiting S100A4.
Immunity, inflammation and disease (2023-02-18)
Min Liu, Shuang Liu, Fajiu Li, Chenghong Li, Shi Chen, Xiaoyan Gao, Xiaojiang Wang
ABSTRACT
Asthma is a chronic respiratory disease with an increasing incidence every year. microRNAs (miRNAs) have been demonstrated to have implications for asthma. However, limited information is available regarding the effect of miR-124-3p on this disease. Therefore, this study aimed to explore the possible effects of miR-124-3p and S100A4 on inflammation and epithelial-mesenchymal transition (EMT) in asthma using mouse models. Ovalbumin was used to induce asthmatic mouse models. Lung injury in mouse models was assessed, and the bronchoalveolar lavage fluid of mice was collected to determine the number of eosinophilic granulocytes and assess inflammation. The expression levels of miR-124-3p, S100A4, E-cadherin, N-cadherin, Snail1, vimentin, and TGF-β1/Smad2 signaling pathway-related proteins were measured using reverse transcription-quantitative polymerase chain reaction (RT-qPCR) and western blot analysis. In vitro experiments, cells were transfected with miR-124-3p mimics or inhibitors to test the expression of S100A4 by RT-qPCR and western blot analysis, and the mutual binding of miR-124-3p and S100A4 was validated by dual-luciferase reporter gene assay. Overexpression of miR-124-3p or inhibition of S100A4 expression attenuated bronchial mucus secretion and collagenous fibers and suppressed inflammatory cell infiltration. Additionally, upon miR-124-3p overexpression or S100A4 suppression, eosinophilic granulocytes were decreased, interleukin-4 (IL-4) and IL-13 expression levels were reduced in the bronchoalveolar lavage fluid, serum total IgE level was reduced, and the TGF-β1/Smad2 signaling pathway was suppressed. Mechanically, a dual-luciferase reporter gene assay verified the binding relationship between miR-124-3p and S100A4. miR-124-3p can negatively target S100A4 to attenuate inflammation in asthmatic mouse models by suppressing the EMT process and the TGF-β/smad2 signaling pathway.
Sigma-Aldrich
Ovalbumin (323-339) (chicken, Japanese quail)