MilliporeSigma
  • Frontline Science: Wnt/β-catenin pathway promotes early engraftment of fetal hematopoietic stem/progenitor cells.
Frontline Science: Wnt/β-catenin pathway promotes early engraftment of fetal hematopoietic stem/progenitor cells.
Journal of leukocyte biology (2018-01-19)
Edward O Kwarteng, Roxann Hétu-Arbour, Krista M Heinonen
ABSTRACT
The switch from fetal to adult hematopoietic stem/progenitor cells (HSPCs) is associated with profound changes in several genetic programs. Although HSPC ageing corresponds to alterations in Wnt signaling, relatively little is known about the relative roles of different Wnt signaling pathways in HSPC ontogeny. We hypothesized that proliferating fetal HSPCs would be more dependent on canonical β-catenin-dependent Wnt signaling when compared to quiescent adult bone marrow HSPCs. We have compared here Wnt signaling activities in murine fetal and adult HSPCs and demonstrate a shift from Wnt/β-catenin-dependent signaling in fetal liver HSPCs to more predominantly noncanonical Wnt/polarity signaling in adult HSPCs. β-Catenin was selectively required for fetal HSPC competitiveness shortly after transplant, and protected cells from oxidative stress. Our results emphasize the complexity of Wnt signaling dynamics in HSPC maintenance and function.
Sigma-Aldrich
Anti-Cdc42 Antibody, from rabbit