MilliporeSigma
  • Generation and Analysis of hTERT-RPE1 VPS54 Knock-Out and Rescued Cell Lines.
Generation and Analysis of hTERT-RPE1 VPS54 Knock-Out and Rescued Cell Lines.
Methods in molecular biology (Clifton, N.J.) (2022-12-14)
Amrita Khakurel, Tetyana Kudlyk, Vladimir V Lupashin
ABSTRACT
The Golgi-associated retrograde protein (GARP) complex is proposed to tether endosome-derived transport vesicles, but the exact function and mechanism of GARP action are not completely understood. To uncover the GARP function in human cells, we employ CRISPR/Cas9 strategy and knock out (KO) the unique VPS54 subunit of the GARP complex. In this chapter, we describe the detailed method of generating CRISPR/Cas9-mediated VPS54-KO in hTERT-RPE1 cells, rescue of resulting KO cells with stable near-endogenous expression of myc-tagged VPS54, and validation of KO and rescued (KO-R) cells using Western blot and immunofluorescence approaches. This approach is helpful in uncovering new functions of the GARP and other vesicle tethering complexes.
Sigma-Aldrich
Monoclonal Anti-Cathepsin D antibody produced in mouse, clone CTD-19, ascites fluid