MilliporeSigma
  • Methods to Measure MDSC Immune Suppressive Activity In Vitro and In Vivo.
Methods to Measure MDSC Immune Suppressive Activity In Vitro and In Vivo.
Current protocols in immunology (2018-10-12)
Samantha Solito, Laura Pinton, Francesco De Sanctis, Stefano Ugel, Vincenzo Bronte, Susanna Mandruzzato, Ilaria Marigo
ABSTRACT
This unit presents methods to assess the immunosuppressive properties of immunoregulatory cells of myeloid origin, such as myeloid-derived suppressor cells (MDSCs), both in vitro and in vivo in mice, as well as in biological samples from cancer patients. These methods could be adapted to test the impact of different suppressive populations on T cell activation, proliferation, and cytotoxic activity; moreover, they could be useful to assess the influence exerted by genetic modifications, chemical inhibitors, and drugs on immune suppressive pathways © 2018 by John Wiley & Sons, Inc.
Sigma-Aldrich
Bovine Serum Albumin, cold ethanol fraction, pH 5.2, ≥96%
Pricing and availability is not currently available.