MilliporeSigma
  • The Condylar Effects of Mesenchymal Stem Cells, Low-Level Laser Therapy and Grape Seed Extract on Functional Mandibular Advancement of the Rat Mandible.
The Condylar Effects of Mesenchymal Stem Cells, Low-Level Laser Therapy and Grape Seed Extract on Functional Mandibular Advancement of the Rat Mandible.
Turkish journal of orthodontics (2023-06-22)
Merve Nur Eğlenen, Mehmet İbrahim Tuğlu, Işıl Aydemir, Ayşegül Güleç
ABSTRACT
Functional treatment of Class II malocclusion is expected to lead to adaptation in the condyle. This study aimed to evaluate the effects of adipose tissue-derived mesenchymal stem cells (ADMSCs), low-level laser therapy (LLLT), and grape-seed extract (GSE) on condylar growth after functional mandibular advancement. Forty-five rats were randomly divided into 8 groups. Functional appliances were applied to all groups (n=6) except the control group (n=3). One group was treated with appliances only; the other six groups received various combinations of ADMSCs, LLLT, and GSE. Analyses for new osteoblasts and new bone formation, vascular endothelial growth factor, and Type II collagen were performed on condylar tissues, after an experimental period of four weeks. The quantitative data obtained from the results of the experiments were evaluated by H-score and analyzed using One-Way ANOVA by Tukey-Kramer multiple comparisons test (p≤0.05). Levels of all investigated parameters increased in all groups (p≤0.05). The highest increases were achieved by a combined application of functional appliance, ADMSCs, LLLT and GSE (p≤0.05). Single LLLT administrations or single GSE applications did not create a statistical difference from appliance alone (p>0.05). A positive effect of ADMSCs or LLLT on osteoblast formation, neovascularization, and Type II collagen level was apparent (p≤0.05), however, neither affected new bone formation (p>0.05). This study shows that ADMSCs with LLLT and GSE applications provide differing levels of new osteoblast and bone formation, new vascular formation, and Type II collagen formation in rat condyles after functional mandibular advancement.
Sigma-Aldrich
Anti-Vascular Endothelial Growth Factor antibody produced in goat, affinity isolated antibody, lyophilized powder