MilliporeSigma
  • Modified Fish Gelatin as an Alternative to Mammalian Gelatin in Modern Food Technologies.
Modified Fish Gelatin as an Alternative to Mammalian Gelatin in Modern Food Technologies.
Polymers (2020-12-24)
Svetlana R Derkach, Nikolay G Voron'ko, Yuliya A Kuchina, Daria S Kolotova
ABSTRACT
This review considers the main properties of fish gelatin that determine its use in food technologies. A comparative analysis of the amino acid composition of gelatin from cold-water and warm-water fish species, in comparison with gelatin from mammals, which is traditionally used in the food industry, is presented. Fish gelatin is characterized by a reduced content of proline and hydroxyproline which are responsible for the formation of collagen-like triple helices. For this reason, fish gelatin gels are less durable and have lower gelation and melting temperatures than mammalian gelatin. These properties impose significant restrictions on the use of fish gelatin in the technology of gelled food as an alternative to porcine and bovine gelatin. This problem can be solved by modifying the functional characteristics of fish gelatin by adding natural ionic polysaccharides, which, under certain conditions, are capable of forming polyelectrolyte complexes with gelatin, creating additional nodes in the spatial network of the gel.
Sigma-Aldrich
Gelatin from cold water fish skin, low endotoxin