MilliporeSigma
Tumour mutations in long noncoding RNAs enhance cell fitness.
Nature communications (2023-06-09)
Roberta Esposito, Andrés Lanzós, Tina Uroda, Sunandini Ramnarayanan, Isabel Büchi, Taisia Polidori, Hugo Guillen-Ramirez, Ante Mihaljevic, Bernard Mefi Merlin, Lia Mela, Eugenio Zoni, Lusine Hovhannisyan, Finn McCluggage, Matúš Medo, Giulia Basile, Dominik F Meise, Sandra Zwyssig, Corina Wenger, Kyriakos Schwarz, Adrienne Vancura, Núria Bosch-Guiteras, Álvaro Andrades, Ai Ming Tham, Michaela Roemmele, Pedro P Medina, Adrian F Ochsenbein, Carsten Riether, Marianna Kruithof-de Julio, Yitzhak Zimmer, Michaela Medová, Deborah Stroka, Archa Fox, Rory Johnson
ABSTRACT
Long noncoding RNAs (lncRNAs) are linked to cancer via pathogenic changes in their expression levels. Yet, it remains unclear whether lncRNAs can also impact tumour cell fitness via function-altering somatic "driver" mutations. To search for such driver-lncRNAs, we here perform a genome-wide analysis of fitness-altering single nucleotide variants (SNVs) across a cohort of 2583 primary and 3527 metastatic tumours. The resulting 54 mutated and positively-selected lncRNAs are significantly enriched for previously-reported cancer genes and a range of clinical and genomic features. A number of these lncRNAs promote tumour cell proliferation when overexpressed in in vitro models. Our results also highlight a dense SNV hotspot in the widely-studied NEAT1 oncogene. To directly evaluate the functional significance of NEAT1 SNVs, we use in cellulo mutagenesis to introduce tumour-like mutations in the gene and observe a significant and reproducible increase in cell fitness, both in vitro and in a mouse model. Mechanistic studies reveal that SNVs remodel the NEAT1 ribonucleoprotein and boost subnuclear paraspeckles. In summary, this work demonstrates the utility of driver analysis for mapping cancer-promoting lncRNAs, and provides experimental evidence that somatic mutations can act through lncRNAs to enhance pathological cancer cell fitness.
Sigma-Aldrich
Anti-SREK1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution