MilliporeSigma
  • Adrenergic α2 receptors are implicated in seizure-induced respiratory arrest in DBA/1 mice.
Adrenergic α2 receptors are implicated in seizure-induced respiratory arrest in DBA/1 mice.
Life sciences (2021-08-31)
Rui Zhang, Zheren Tan, Jianguo Niu, Hua-Jun Feng
ABSTRACT
Sudden unexpected death in epilepsy (SUDEP) is a serious and underestimated public health burden. Both clinical and animal studies show that seizure-induced respiratory arrest (S-IRA) is the primary cause of death in SUDEP. Our previous studies demonstrated that atomoxetine, a norepinephrine reuptake inhibitor (NRI), suppresses S-IRA in DBA/1 mice, suggesting that noradrenergic neurotransmission modulates S-IRA. However, it remains unclear which adrenoceptors are implicated in S-IRA in DBA/1 mice. Naïve DBA/1 mice exhibit a low incidence of S-IRA, but after primed by acoustic stimulation, they become consistently susceptible to S-IRA. Atomoxetine, adrenoceptor agonists, antagonists or vehicle was intraperitoneally (i.p.) administered alone or in combination, and the effects of drug treatments on S-IRA incidence and seizure behaviors were examined. The incidence of S-IRA in primed DBA/1 mice was significantly reduced by clonidine, an α2 adrenoceptor agonist, as compared with that of the vehicle control. However, compared with the vehicle control, S-IRA was not altered by cirazoline, an α1 agonist. Consistent with previous reports, atomoxetine reduced S-IRA in primed DBA/1 mice. The suppressing effect of atomoxetine on S-IRA was prevented by injection of an α2 adrenoceptor antagonist, yohimbine or atipamezole, but not by prazosin, an α1 antagonist. Administration of α1 or α2 antagonists alone did not promote the incidence of S-IRA in nonprimed DBA/1 mice. These data demonstrate that noradrenergic neurotransmission modulates S-IRA predominantly via α2 adrenoceptors in DBA/1 mice, indicating that selective activation of α2 adrenoceptors can potentially prevent SUDEP.
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)