MilliporeSigma
  • Discovery and In Vitro Characterization of BAY 2686013, an Allosteric Small Molecule Antagonist of the Human Pituitary Adenylate Cyclase-Activating Polypeptide Receptor.
Discovery and In Vitro Characterization of BAY 2686013, an Allosteric Small Molecule Antagonist of the Human Pituitary Adenylate Cyclase-Activating Polypeptide Receptor.
Molecular pharmacology (2023-06-23)
Gernot Langer, John Scott, Christoffer Lind, Christiane Otto, Ulrich Bothe, Alexis Laux-Biehlmann, Jörg Müller, Beau le Roy, Horst Irlbacher, Katrin Nowak-Reppel, Anne Schlüter, Adam J Davenport, Mark Slack, Stefan Bäurle
ABSTRACT
The human pituitary adenylate cyclase-activating polypeptide receptor (hPAC1-R), a class B G-protein-coupled receptor (GPCR) identified almost 30 years ago, represents an important pharmacological target in the areas of neuroscience, oncology, and immunology. Despite interest in this target, only a very limited number of small molecule modulators have been reported for this receptor. We herein describe the results of a drug discovery program aiming for the identification of a potent and selective hPAC1-R antagonist. An initial high-throughput screening (HTS) screen of 3.05 million compounds originating from the Bayer screening library failed to identify any tractable hits. A second, completely revised screen using native human embryonic kidney (HEK)293 cells yielded a small number of hits exhibiting antagonistic properties (4.2 million compounds screened). BAY 2686013 (1) emerged as a promising compound showing selective antagonistic activity in the submicromolar potency range. In-depth characterization supported the hypothesis that BAY 2686013 blocks receptor activity in a noncompetitive manner. Preclinical, pharmacokinetic profiling indicates that BAY 2686013 is a valuable tool compound for better understanding the signaling and function of hPAC1-R. SIGNIFICANCE STATEMENT: Although the human pituitary adenylate cyclase-activating polypeptide receptor (hPAC1-R) is of major significance as a therapeutic target with a well documented role in pain signaling, only a very limited number of small-molecule (SMOL) compounds are known to modulate its activity. We identified and thoroughly characterized a novel, potent, and selective SMOL antagonist of hPAC1-R (acting in an allosteric manner). These characteristics make BAY 2686013 an ideal tool for further studies.
Sigma-Aldrich
BAY 2686013, ≥98% (HPLC)