MilliporeSigma
Cooperative Cu/Pd-catalyzed borocarbonylation of ethylene.
Chemical communications (Cambridge, England) (2022-10-14)
Yang Yuan, Jian-Xing Xu, Xiao-Feng Wu
ABSTRACT
We report here a cooperative Cu/Pd-catalyzed multi-component borocarbonylation of ethylene with aryl iodides. A variety of synthetically useful β-boryl ketones were assembled from the most basic C1 (CO) and C2 (ethylene) building blocks in good yields.
Sigma-Aldrich
DPPF ChemBeads