MilliporeSigma
  • NS1619 regulates the expression of caveolin-1 protein in a time-dependent manner via ROS/PI3K/PKB/FoxO1 signaling pathway in brain tumor microvascular endothelial cells.
NS1619 regulates the expression of caveolin-1 protein in a time-dependent manner via ROS/PI3K/PKB/FoxO1 signaling pathway in brain tumor microvascular endothelial cells.
Journal of the neurological sciences (2016-09-23)
Rui-Ping Cai, Yi-Xue Xue, Jian Huang, Jin-Hui Wang, Jia-Hong Wang, Song-Yan Zhao, Ting-Ting Guan, Zhou Zhang, Yan-Ting Gu
ABSTRACT
NS1619, a calcium-activated potassium channel (Kca channel) activator, can selectively and time-dependently accelerate the formation of transport vesicles in both the brain tumor capillary endothelium and tumor cells within 15min of treatment and then increase the permeability of the blood-brain tumor barrier (BTB). However, the mechanism involved is still under investigation. Using a rat brain glioma (C6) model, the expression of caveolin-1, FoxO1 and p-FoxO1 protein were examined at different time points after intracarotid infusion of NS1619 at a dose of 30μg/kg/min. Internalization of Cholera toxin subunit (CTB) labeled fluorescently was monitored by flow cytometry. The expression of caveolin-1 and FoxO1 protein at tumor microvessels was enhanced and caveolae-mediated CTB endocytosis was increased by NS1619 infusion for 15min. Compared with the 15min group, the expression of caveolin-1 protein was significantly decreased and the level of phosphorylation of FoxO1 was significantly increased in the NS1619 2h group. In addition, inhibitors of reactive oxygen species (ROS) or PI3K or PKB significantly attenuated the level of FoxO1 phosphorylation and also increased the expression of caveolin-1 protein in Human Brain Microvascular Endothelial Cells (HBMECs) cocultured with human glioma cells (U87) 2h after NS1619 treatment. This led to the conclusion that NS1619-mediated transport vesicle increase is, at least partly, related to the ROS/PI3K/PKB/FoxO1 signaling pathway.
Sigma-Aldrich
NS 1619, powder