MilliporeSigma
  • Investigation of Staphylococcus aureus Biofilm-Associated Toxin as a Potential Squamous Cell Carcinoma Therapeutic.
Investigation of Staphylococcus aureus Biofilm-Associated Toxin as a Potential Squamous Cell Carcinoma Therapeutic.
Microorganisms (2024-02-24)
Zi Xin Ong, Bavani Kannan, Anthony R J Phillips, David L Becker
ABSTRACT
Cancer therapies developed using bacteria and their components have been around since the 19th century. Compared to traditional cancer treatments, the use of bacteria-derived compounds as cancer therapeutics could offer a higher degree of specificity, with minimal off-target effects. Here, we explored the use of soluble bacteria-derived toxins as a potential squamous cell carcinoma (SCC) therapeutic. We optimized a protocol to generate Staphylococcus aureus biofilm-conditioned media (BCM), where soluble bacterial products enriched in the development of biofilms were isolated from a bacterial culture and applied to SCC cell lines. Bioactive components of S. aureus ATCC 29213 (SA29213) BCM display selective toxicity towards cancerous human skin SCC-12 at low doses, while non-cancerous human keratinocyte HaCaT and fibroblast BJ-5ta are minimally affected. SA29213 BCM treatment causes DNA damage to SCC-12 and initiates Caspase 3-dependent-regulated cell death. The use of the novel SA29213 bursa aurealis transposon mutant library led to the identification of S. aureus alpha hemolysin as the main bioactive compound responsible for the observed SCC-12-specific toxicity. The antibody neutralisation of Hla eradicates the cytotoxicity of SA29213 BCM towards SCC-12. Hla displays high SCC-12-specific toxicity, which is exerted primarily through Hla-ADAM10 interaction, Hla oligomerisation, and pore formation. The high target specificity and potential to cause cell death in a controlled manner highlight SA29213 Hla as a good candidate as an alternative SCC therapeutic.
Sigma-Aldrich
Necrostatin-1, ≥98% (HPLC)
Sigma-Aldrich
Anti-Staphylococcal α-Toxin antibody produced in rabbit, whole antiserum
Sigma-Aldrich
Ferrostatin-1, ≥95% (HPLC)