MilliporeSigma
  • A Detailed Protocol for Constructing a Human Single-Chain Variable Fragment (scFv) Library and Downstream Screening via Phage Display.
A Detailed Protocol for Constructing a Human Single-Chain Variable Fragment (scFv) Library and Downstream Screening via Phage Display.
Methods and protocols (2024-02-23)
Ziyi Liu, Dokyun Kim, Seokmin Kang, Jae U Jung
ABSTRACT
The development of monoclonal antibodies (mAbs) represents a significant milestone in both basic research and clinical applications due to their target specificity and versatility in therapeutic and diagnostic applications. The innovative strategy of mAb screening, utilizing phage display, facilitates the in vitro screening of antibodies with high affinity to target antigens. The single-chain variable fragment (scFv) is a subset of mAb derivatives, known for its high binding affinity and smaller size-just one-third of that of human IgG. This report outlines a detailed and comprehensive procedure for constructing a scFv phagemid library derived from human patients, followed by screening via phage display affinity selection. The protocol utilizes 348 primer combinations spanning the entire human antibody repertoire to minimize sequence bias and maintain library diversity during polymerase chain reaction (PCR) for scFv generation, resulting in a library size greater than 1 × 108. Furthermore, we describe a high-throughput phage display screening protocol using enzyme-linked immunosorbent assay (ELISA) to evaluate more than 1200 scFv candidates. The generation of a highly diverse scFv library, coupled with the implementation of a phage display screening methodology, is expected to provide a valuable resource for researchers in pursuit of scFvs with high affinity for target antigens, thus advancing both research and clinical endeavors.
Sigma-Aldrich
AEBSF, Hydrochloride, AEBSF, CAS 30827-99-7, is a specific irreversible inhibitor of chymotrypsin, kallikrein, plasmin, thrombin, trypsin, and related thrombolytic enzymes. A nontoxic alternative to PMSF and DFP.