MilliporeSigma
  • RNF8 ubiquitylation of XRN2 facilitates R-loop resolution and restrains genomic instability in BRCA1 mutant cells.
RNF8 ubiquitylation of XRN2 facilitates R-loop resolution and restrains genomic instability in BRCA1 mutant cells.
Nucleic acids research (2023-09-12)
Rehna Krishnan, Mariah Lapierre, Brandon Gautreau, Kevin C J Nixon, Samah El Ghamrasni, Parasvi S Patel, Jun Hao, V Talya Yerlici, Kiran Kumar Naidu Guturi, Jonathan St-Germain, Francesca Mateo, Amine Saad, Arash Algouneh, Rebecca Earnshaw, Duan Shili, Alma Seitova, Joshua Miller, Negin Khosraviani, Adam Penn, Brandon Ho, Otto Sanchez, M Prakash Hande, Jean-Yves Masson, Grant W Brown, Moulay Alaoui-Jamali, John J Reynolds, Cheryl Arrowsmith, Brian Raught, Miguel A Pujana, Karim Mekhail, Grant S Stewart, Anne Hakem, Razqallah Hakem
ABSTRACT
Breast cancer linked with BRCA1/2 mutations commonly recur and resist current therapies, including PARP inhibitors. Given the lack of effective targeted therapies for BRCA1-mutant cancers, we sought to identify novel targets to selectively kill these cancers. Here, we report that loss of RNF8 significantly protects Brca1-mutant mice against mammary tumorigenesis. RNF8 deficiency in human BRCA1-mutant breast cancer cells was found to promote R-loop accumulation and replication fork instability, leading to increased DNA damage, senescence, and synthetic lethality. Mechanistically, RNF8 interacts with XRN2, which is crucial for transcription termination and R-loop resolution. We report that RNF8 ubiquitylates XRN2 to facilitate its recruitment to R-loop-prone genomic loci and that RNF8 deficiency in BRCA1-mutant breast cancer cells decreases XRN2 occupancy at R-loop-prone sites, thereby promoting R-loop accumulation, transcription-replication collisions, excessive genomic instability, and cancer cell death. Collectively, our work identifies a synthetic lethal interaction between RNF8 and BRCA1, which is mediated by a pathological accumulation of R-loops.
Sigma-Aldrich
Actinomycin D, from Streptomyces sp., suitable for cell culture, ≥95%
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-phospho-Histone H2A.X (Ser139) Antibody, clone JBW301, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Anti-phospho-H2A.X (Ser139) Antibody, Upstate®, from rabbit