MilliporeSigma
  • Radial astrocyte synchronization modulates the visual system during behavioral-state transitions.
Radial astrocyte synchronization modulates the visual system during behavioral-state transitions.
Neuron (2023-10-21)
Alejandro Uribe-Arias, Rotem Rozenblat, Ehud Vinepinsky, Emiliano Marachlian, Anirudh Kulkarni, David Zada, Martin Privat, Diego Topsakalian, Sarah Charpy, Virginie Candat, Sarah Nourin, Lior Appelbaum, Germán Sumbre
ABSTRACT
Glial cells support the function of neurons. Recent evidence shows that astrocytes are also involved in brain computations. To explore whether and how their excitable nature affects brain computations and motor behaviors, we used two-photon Ca2+ imaging of zebrafish larvae expressing GCaMP in both neurons and radial astrocytes (RAs). We found that in the optic tectum, RAs synchronize their Ca2+ transients immediately after the end of an escape behavior. Using optogenetics, ablations, and a genetically encoded norepinephrine sensor, we observed that RA synchronous Ca2+ events are mediated by the locus coeruleus (LC)-norepinephrine circuit. RA synchronization did not induce direct excitation or inhibition of tectal neurons. Nevertheless, it modulated the direction selectivity and the long-distance functional correlations among neurons. This mechanism supports freezing behavior following a switch to an alerted state. These results show that LC-mediated neuro-glial interactions modulate the visual system during transitions between behavioral states.
Sigma-Aldrich
Prazosin hydrochloride, ≥99.0% (HPLC)