MilliporeSigma
  • GPhos Ligand Enables Production of Chiral N-Arylamines in a Telescoped Transaminase-Buchwald-Hartwig Amination Cascade in the Presence of Excess Amine Donor.
GPhos Ligand Enables Production of Chiral N-Arylamines in a Telescoped Transaminase-Buchwald-Hartwig Amination Cascade in the Presence of Excess Amine Donor.
Chemistry (Weinheim an der Bergstrasse, Germany) (2021-09-30)
Christian M Heckmann, Francesca Paradisi
ABSTRACT
The combination of biocatalysis and chemocatalysis can be more powerful than either technique alone. However, combining the two is challenging due to typically very different reaction conditions. Herein, chiral N-aryl amines, key features of many active pharmaceutical ingredients, are accessed in excellent enantioselectivity (typically>99.5 % ee) by combining transaminases with the Buchwald-Hartwig amination. By employing a bi-phasic buffer-toluene system as well as the ligand GPhos, the telescoped cascade proceeded with up to 89 % overall conversion in the presence of excess alanine. No coupling to alanine was observed.
Sigma-Aldrich
GPhos Pd G3 ChemBeads