Merck
  • Effect of different purification techniques on the characteristics of heteropolysaccharide-protein biopolymer from durian (Durio zibethinus) seed.
Effect of different purification techniques on the characteristics of heteropolysaccharide-protein biopolymer from durian (Durio zibethinus) seed.
Molecules (Basel, Switzerland) (2012-09-12)
Bahareh Tabatabaee Amid, Hamed Mirhosseini
ABSTRACT
Natural biopolymers from plant sources contain many impurities (e.g., fat, protein, fiber, natural pigment and endogenous enzymes), therefore, an efficient purification process is recommended to minimize these impurities and consequently improve the functional properties of the biopolymer. The main objective of the present study was to investigate the effect of different purification techniques on the yield, protein content, solubility, water- and oil-holding capacity of a heteropolysaccharide-protein biopolymer obtained from durian seed. Four different purification methods using different chemicals and solvents (i.e., A (isopropanol and ethanol), B (isopropanol and acetone), C (saturated barium hydroxide), and D (Fehling solution)] to liberate the purified biopolymer from its crude form were compared. In most cases, the purification process significantly (p < 0.05) improved the physicochemical properties of heteropolysaccharide-protein biopolymer from durian fruit seed. The present work showed that the precipitation using isopropanol and acetone (Method B) resulted in the highest purification yield among all the tested purification techniques. The precipitation using saturated barium hydroxide (Method C) led to induce the highest solubility and relatively high capacity of water absorption. The current study reveals that the precipitation using Fehling solution (Method D) most efficiently eliminates the protein fraction, thus providing more pure biopolymer suitable for biological applications.
Sigma-Aldrich
Barium hydroxide solution, 0.3 N