MilliporeSigma
  • A novel core protein as well as polymorphic epithelial mucin carry peanut agglutinin binding sites in human gastric carcinoma cells: sequence analysis and examination of gene expression.
A novel core protein as well as polymorphic epithelial mucin carry peanut agglutinin binding sites in human gastric carcinoma cells: sequence analysis and examination of gene expression.
Journal of biochemistry (1992-11-01)
Y Masuzawa, T Miyauchi, M Hamanoue, S Ando, J Yoshida, S Takao, H Shimazu, M Adachi, T Muramatsu
ABSTRACT
The peanut agglutinin (PNA)-binding site is protein-bound Gal beta 1-->3GalNAc, and is a tumor-associated carbohydrate marker expressed in many human carcinomas. PNA-binding glycoproteins isolated from KATO-III human gastric carcinoma cells were deglycosylated by trifluoromethanesulfonic acid, and rabbit antibodies against the core proteins were used to screen a lambda gt11 expression library constructed from these cells. Two different core proteins were identified by this approach. One was polymorphic epithelial mucin (PEM), initially found in breast carcinomas. PEM mRNA was expressed in normal tissues of the stomach, colon, and lung, but not in the small intestine, thyroid, and spleen. High levels of PEM mRNA were detected in some nude mouse-transplanted carcinomas, i.e. colorectal, pancreatic, stomach, and lung carcinomas. The other core protein was a novel one called MGC-24, which has a molecular mass of 24 kDa, is rich in hydroxyl amino acids and cysteine, and lacks repeating motifs. The mature MGC-24 glycoprotein behaved as a high-molecular-mass one upon SDS-polyacrylamide gel electrophoresis even after neuraminidase treatment. Treatment with endo-alpha-N-acetylgalactosaminidase in the absence of neuraminidase significantly changed the staining pattern by anti-MGC-24, confirming that MGC-24 carried PNA-binding sites. MGC-24 mRNA was intensely expressed in normal tissues of the colon, small intestine and thyroid, and in some nude mouse-transplanted colorectal and pancreatic adenocarcinomas.
Sigma-Aldrich
CD164 human, recombinant, expressed in E. coli, 0.5 mg protein/mL
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
50 μg
Usually ships in 3 weeks.
$258.00