MilliporeSigma
  • Rescue of fragile X syndrome phenotypes in Fmr1 KO mice by the small-molecule PAK inhibitor FRAX486.
Rescue of fragile X syndrome phenotypes in Fmr1 KO mice by the small-molecule PAK inhibitor FRAX486.
Proceedings of the National Academy of Sciences of the United States of America (2013-03-20)
Bridget M Dolan, Sergio G Duron, David A Campbell, Benedikt Vollrath, B S Shankaranarayana Rao, Hui-Yeon Ko, Gregory G Lin, Arvind Govindarajan, Se-Young Choi, Susumu Tonegawa
ABSTRACT
Fragile X syndrome (FXS) is the most common inherited form of autism and intellectual disability and is caused by the silencing of a single gene, fragile X mental retardation 1 (Fmr1). The Fmr1 KO mouse displays phenotypes similar to symptoms in the human condition--including hyperactivity, repetitive behaviors, and seizures--as well as analogous abnormalities in the density of dendritic spines. Here we take a hypothesis-driven, mechanism-based approach to the search for an effective therapy for FXS. We hypothesize that a treatment that rescues the dendritic spine defect in Fmr1 KO mice may also ameliorate autism-like behavioral symptoms. Thus, we targeted a protein that regulates spines through modulation of actin cytoskeleton dynamics: p21-activated kinase (PAK). Our results demonstrate that a potent small molecule inhibitor of group I PAKs reverses dendritic spine phenotypes in Fmr1 KO mice. Moreover, this PAK inhibitor--which we call FRAX486--also rescues seizures and behavioral abnormalities such as hyperactivity and repetitive movements, thereby supporting the hypothesis that a drug treatment that reverses the spine abnormalities can also treat neurological and behavioral symptoms. Finally, a single administration of FRAX486 is sufficient to rescue all of these phenotypes in adult Fmr1 KO mice, demonstrating the potential for rapid, postdiagnostic therapy in adults with FXS.
Sigma-Aldrich
(2-Hydroxypropyl)-β-cyclodextrin solution
Sigma-Aldrich
FRAX486, ≥98% (HPLC)