MilliporeSigma
  • Benzoylated ethyl 1-thioglycosides: direct preparation from per-O-benzoylated sugars.
Benzoylated ethyl 1-thioglycosides: direct preparation from per-O-benzoylated sugars.
Carbohydrate research (2012-06-29)
Deepak Sail, Pavol Kováč
ABSTRACT
D-Glucose, lactose, maltose, and melibiose were benzoylated with Bz(2)O-Et(3)N reagent to give fully benzoylated β products. Under the same conditions, D-mannose produced a mixture where the β-benzoate predominated. Treatment of the foregoing compounds with EtSH at slightly elevated temperature (50-60°C) in the presence of BF(3)·Et(2)O as a promoter gave the corresponding ethyl 1-thio glycosides in high yields. The α-products predominated in all cases in the anomeric mixtures formed. Individual products of all reactions were isolated by chromatography, they were obtained in analytically pure state, and were fully characterized by (1)H and (13)C NMR data and physical constants.
Sigma-Aldrich
Ethanethiol, 97%
Sigma-Aldrich
Ethanethiol, ≥97%
Sigma-Aldrich
Melibiose, ≥98% (HPLC)
Sigma-Aldrich
Sodium ethanethiolate, technical, ~90% (RT)
Sigma-Aldrich
Sodium ethanethiolate, technical grade, 80%
Sigma-Aldrich
Boron trifluoride diethyl etherate, for synthesis
Sigma-Aldrich
Boron trifluoride diethyl etherate, purified by redistillation, ≥46.5% BF3 basis