MilliporeSigma
  • ERBB3 (HER3) is a key sensor in the regulation of ERBB-mediated signaling in both low and high ERBB2 (HER2) expressing cancer cells.
ERBB3 (HER3) is a key sensor in the regulation of ERBB-mediated signaling in both low and high ERBB2 (HER2) expressing cancer cells.
Cancer medicine (2013-01-24)
Byung-Kwon Choi, Xuejun Fan, Hui Deng, Ningyan Zhang, Zhiqiang An
ABSTRACT
Aberrant expression and activation of EGFR and ERBB2 (HER2) have been successfully targeted for cancer therapeutics. Recent evidence from both basic and clinical studies suggests that ERBB3 (HER3) serves as a key activator of downstream signaling through dimerization with other ERBB proteins and plays a critical role in the widespread clinical resistance to EGFR and HER2 targeting cancer therapies. As a result, HER3 is actively pursued as an antibody therapeutic target for cancer. Ligand binding is thought to be a prerequisite for dimerization of HER3 with other ERBB proteins, which results in phosphorylation of its c-terminal tyrosine residues and activation of downstream AKT and MAPK signaling pathways. In this study, we report that an anti-HER2 monoclonal antibody (HER2Mab), which blocks HER2 dimerization with HER3, induces HER3 dimerization with EGFR in both low and high HER2 expressing cancer cells. Treatment of the low HER2 expressing MCF7 cancer cells with HER2Mab promoted cell proliferation and migration in the absence of HER3 ligand stimulation. Follow-up studies revealed that HER2Mab-induced HER3 signaling via EGFR/HER3 dimerization and activation of downstream AKT signaling pathways. These results suggest that equilibrium of dimerization among the ERBB proteins can be perturbed by HER2Mab and HER3 plays a key role in sensing the perturbation.
Sigma-Aldrich
Duolink® In Situ Wash Buffers, Fluorescence
Sigma-Aldrich
Duolink® In Situ Detection Reagents Orange
Sigma-Aldrich
Duolink® In Situ Detection Reagents FarRed
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat PLUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Green
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit MINUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Rabbit PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Mouse MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Microplate Nuclear Stain, Anti-Fade
Sigma-Aldrich
Duolink® In Situ Mounting Medium with DAPI
Sigma-Aldrich
Duolink® In Situ Detection Reagents Red
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Microplate Heat Transfer Block