Merck
  • Evidence for 5-HT7 receptors mediating relaxation of human colonic circular smooth muscle.
Evidence for 5-HT7 receptors mediating relaxation of human colonic circular smooth muscle.
British journal of pharmacology (1999-11-11)
N H Prins, M R Briejer, P J Van Bergen, L M Akkermans, J A Schuurkes
ABSTRACT
5-HT4 receptors mediate relaxation of human colon circular muscle. However, after 5-HT4 receptor blockade (SB 204070 10 nM), 5-HT still induced a relaxation (pEC50 6.3). 5-HT4 receptors were sufficiently blocked, as the curves to 5-HT obtained in the presence of 10 and 100 nM SB 204070 were indistinguishable. This 5-HT-induced relaxation was tetrodotoxin-insensitive, indicative of a smooth muscle relaxant 5-HT receptor. This, and the rank order of potency (5-CT=5-MeOT=5-HT) suggested involvement of 5-HT1 or 5-HT7 receptors. Mesulergine, a 5-HT7 receptor antagonist at nanomolar concentrations, and a 5-HT1 receptor antagonist at micromolar concentrations, competitively antagonized the 5-HT-induced relaxation (pKB 8.3) and antagonized the relaxation to 5-CT. Methysergide antagonized the 5-HT-induced relaxation (pA2 7.6). It is concluded that the profile of the smooth muscle inhibitory 5-HT receptor resembles that of the 5-HT7 receptor. These data provide the first evidence for functional human 5-HT7 receptors.
Sigma-Aldrich
SB-204070 hydrochloride, solid, ≥97% (HPLC)