MilliporeSigma
  • Transglycosylation and reverse hydrolysis reactions of endoglycoceramidase from the jellyfish, Cyanea nozakii.
Transglycosylation and reverse hydrolysis reactions of endoglycoceramidase from the jellyfish, Cyanea nozakii.
Journal of biochemistry (2001-08-02)
Y Horibata, H Higashi, M Ito
ABSTRACT
Endoglycoceramidase (EGCase: EC 3.2.1.123) is an enzyme capable of cleaving the glycosidic linkage between oligosaccharides and ceramides in various glycosphingolipids. We report here transglycosylation and reverse hydrolysis reactions of EGCase from the jellyfish Cynaea nozakii. Various alkyl-GM1 oligosaccharides (alkyl-II(3)NeuAcGgOse4) were synthesized when GM1 ganglioside was treated with the EGCase in the presence of 1-alkanols. Among various 1-alkanols tested, methanol was found to be the most preferential acceptor, followed by 1-hexanol and 1-pentanol. GM1 was the best donor, followed by GD1b and GT1b, when methanol was used as an acceptor. However, neither globoside nor glucosylceramide was utilized by the enzyme as a donor substrate. The enzyme transferred oligosaccharides from various glycosphingolipids to NBD-ceramide, a fluorescent ceramide, producing NBD-labeled glycosphingolipids. In addition to the transglycosylation reaction, the enzyme catalyzed the reverse hydrolysis reaction; lactose was condensed to ceramide to generate lactosylceramide in the presence of the enzyme. These results indicate that the jellyfish enzyme will facilitate the synthesis of various neoglycoconjugates and glycosphingolipids.
Sigma-Aldrich
Endoglycoceramidase II from Rhodococcus sp., aqueous solution
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
100 ea
In Stock
Details...
$84.80