MilliporeSigma
  • Development of pyrF-based genetic system for targeted gene deletion in Clostridium thermocellum and creation of a pta mutant.
Development of pyrF-based genetic system for targeted gene deletion in Clostridium thermocellum and creation of a pta mutant.
Applied and environmental microbiology (2010-08-10)
Shital A Tripathi, Daniel G Olson, D Aaron Argyros, Bethany B Miller, Trisha F Barrett, Daniel M Murphy, Jesse D McCool, Anne K Warner, Vineet B Rajgarhia, Lee R Lynd, David A Hogsett, Nicky C Caiazza
ABSTRACT
We report development of a genetic system for making targeted gene knockouts in Clostridium thermocellum, a thermophilic anaerobic bacterium that rapidly solubilizes cellulose. A toxic uracil analog, 5-fluoroorotic acid (5-FOA), was used to select for deletion of the pyrF gene. The ΔpyrF strain is a uracil auxotroph that could be restored to a prototroph via ectopic expression of pyrF from a plasmid, providing a positive genetic selection. Furthermore, 5-FOA was used to select against plasmid-expressed pyrF, creating a negative selection for plasmid loss. This technology was used to delete a gene involved in organic acid production, namely pta, which encodes the enzyme phosphotransacetylase. The C. thermocellum Δpta strain did not produce acetate. These results are the first examples of targeted homologous recombination and metabolic engineering in C. thermocellum, a microbe that holds an exciting and promising future in the biofuel industry and development of sustainable energy resources.
Sigma-Aldrich
Phosphotransacetylase from Bacillus stearothermophilus, lyophilized powder, ≥3,000 units/mg protein (biuret)