MilliporeSigma
  • Diagnosis and treatment trends in mucopolysaccharidosis I: findings from the MPS I Registry.
Diagnosis and treatment trends in mucopolysaccharidosis I: findings from the MPS I Registry.
European journal of pediatrics (2012-01-12)
Kristin D'Aco, Lisa Underhill, Lakshmi Rangachari, Pamela Arn, Gerald F Cox, Roberto Giugliani, Torayuki Okuyama, Frits Wijburg, Paige Kaplan
ABSTRACT
Our objective was to assess how the diagnosis and treatment of mucopolysaccharidosis I (MPS I) have changed over time. We used data from 891 patients in the MPS I Registry, an international observational database, to analyze ages at symptom onset, diagnosis, treatment initiation, and treatment allocation (hematopoietic stem cell transplantation, enzyme replacement therapy with laronidase, both, or neither) over time for all disease phenotypes (Hurler, Hurler-Scheie, and Scheie syndromes). The interval between diagnosis and treatment has become shorter since laronidase became available in 2003 (gap during 2006-2009: Hurler--0.2 year, Hurler-Scheie--0.5 year, Scheie--1.4 years). However, the age at diagnosis has not decreased for any MPS I phenotype over time, and the interval between symptom onset and treatment initiation remains substantial for both Hurler-Scheie and Scheie patients (gap during 2006-2009, 2.42 and 6.71 years, respectively). Among transplanted patients, an increasing proportion received hematopoietic stem cells from cord blood (34 out of 64 patients by 2009) and was also treated with laronidase (42 out of 45 patients by 2009). Despite the availability of laronidase since 2003, the diagnosis of MPS I is still substantially delayed for patients with Hurler-Scheie and Scheie phenotypes, which can lead to a sub-optimal treatment outcome. Increased awareness of MPS I signs and symptoms by primary care providers and pediatric subspecialists is crucial to initiate early treatment and to improve the quality of life of MPS I patients.
Sigma-Aldrich
α-L-Iduronidase human, recombinant, expressed in mouse NSO cells