Merck
  • Human papillomavirus sperm infection and assisted reproduction: a dangerous hazard with a possible safe solution.
Human papillomavirus sperm infection and assisted reproduction: a dangerous hazard with a possible safe solution.
Human reproduction (Oxford, England) (2012-02-09)
Andrea Garolla, Andrea Lenzi, Giorgio Palù, Damiano Pizzol, Alessandro Bertoldo, Luca De Toni, Carlo Foresta
ABSTRACT
Human papillomavirus (HPV) infection has been demonstrated in the sperm of a large percentage of sexually active males and is associated with an impairment of sperm parameters, with a particular negative impact on sperm motility, suggesting a possible role in male infertility. Conventional sperm selection techniques have a low efficiency in removing HPV. Evaluation of sperm parameters, terminal deoxynucleotidyltransferase-mediated dUTP nick-end labeling test to evaluate DNA fragmentation and fluorescence in situ hybridization or immunohistochemistry for HPV were performed on semen samples from infected patients (n= 22), control subjects (n= 13) and on pooled control sperm samples incubated with HPV16-L1 (HPV capsid), before and after direct swim-up and modified swim-up (with added Heparinase-III). Moreover, cytofluorimetry for HPV detection was performed in pooled sperm pre- and post-incubation with HPV 16-L1 before and after direct and modified swim-up. Statistical analysis was performed with a two-tailed Student's t-test. Direct swim-up reduces the number of HPV-infected sperm by ~24% (P< 0.01), while modified swim-up is able to remove completely HPV DNA both from naturally and artificially infected sperm. Enzymatic treatment with Heparinase-III tended to decrease sperm motility, viability and DNA integrity but the effects were not significant. This study shows that Heparinase-III treatment seems not to affect spermatozoa in vitro and suggests that this treatment should be investigated further as a means of preparing sperm from patients who are infected with HPV in order to reduce the risk of HPV infection when using assisted reproduction techniques.
Sigma-Aldrich
Heparinase III from Flavobacterium heparinum, Lyophilized powder stabilized with approx. 25% (w/w) bovine serum albumin, ≥30 units/mg protein (enzyme + BSA)