Merck
  • Levamisole-induced reduction in seizure threshold: a possible role of nicotinic acetylcholine receptor-mediated pathway.
Levamisole-induced reduction in seizure threshold: a possible role of nicotinic acetylcholine receptor-mediated pathway.
Naunyn-Schmiedeberg's archives of pharmacology (2010-08-10)
Ashish K Rehni, Thakur Gurjeet Singh
ABSTRACT
Levamisole produces seizures in man and is also known to activate the neuronal nicotinic acetylcholine receptors, which are further known to elicit seizure activity. Therefore, the present study has been designed to investigate the role of nicotinic acetylcholine receptor activation in the seizure-inducing effect of levamisole. Levamisole, at the doses of 5, 10 and 20 mg kg(-1), i.p. were used to elicit seizures in mice. Seizures were assessed in terms of the onset time of Straub's tail phenomenon, jerky movements of whole body and convulsions. Additionally, an isobolographic design was used to examine the interaction of systemically administered levamisole and nicotine. Prior administration of 2, 2, 6, 6-tetramethylpiperidin-4-yl heptanoate (TMPH), a selective inhibitor of neuronal nicotinic acetylcholine receptors, markedly attenuated the seizure-inducing effect of levamisole. Moreover, administration of TMPH per se did not produce any behavioural changes in mice. Furthermore, nicotine was observed exert a synergistic interaction with levamisole. Therefore, it may be suggested that levamisole-induced reduction in seizure threshold might be ascribed to the activation of a neuronal nicotinic acetylcholine receptor activation-linked mechanism.
Sigma-Aldrich
TMPH hydrochloride, solid, (Product is pure based on CHN, NMR and MS results)